Merck
  • Controlling neural territory patterning from pluripotency using a systems developmental biology approach.
Controlling neural territory patterning from pluripotency using a systems developmental biology approach.
iScience (2022-04-19)
Katie E Sears, Keerthi Gullapalli, Divya Trivedi, Alexander Mihas, Michael A Bukys, Jan Jensen
ABSTRACT
Successful manufacture of specialized human cells requires process understanding of directed differentiation. Here, we apply high-dimensional Design of Experiments (HD-DoE) methodology to identify critical process parameters (CPPs) that govern neural territory patterning from pluripotency-the first stage toward specification of central nervous system (CNS) cell fates. Using computerized experimental design, 7 developmental signaling pathways were simultaneously perturbed in human pluripotent stem cell culture. Regionally specific genes spanning the anterior-posterior and dorsal-ventral axes of the developing embryo were measured after 3 days and mathematical models describing pathway control were developed using regression analysis. High-dimensional models revealed particular combinations of signaling inputs that induce expression profiles consistent with emerging CNS territories and defined CPPs for anterior and posterior neuroectoderm patterning. The results demonstrate the importance of combinatorial control during neural induction and challenge the use of generic neural induction strategies such as dual-SMAD inhibition, when seeking to specify particular lineages from pluripotency.
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Roche
Insulin, human, recombinant (yeast)
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.
Sigma-Aldrich
A 83-01, ≥98% (HPLC)