Merck
  • Preparation and characterization of activated carbon produced from rice straw by (NH4)2HPO4 activation.
Preparation and characterization of activated carbon produced from rice straw by (NH4)2HPO4 activation.
Bioresource technology (2010-12-15)
Pin Gao, Zhen-hong Liu, Gang Xue, Bin Han, Mei-hua Zhou
ABSTRACT
Effects of different pretreatment protocols in (NH(4))(2)HPO(4) activation of rice straw on porous activated carbon evolution were evaluated. The pore structure, morphology and surface chemistry of obtained activated carbons were investigated by nitrogen adsorption, scanning electron microscopy and Fourier transform infrared spectroscopy. It was found that pretreatment combining impregnation with (NH(4))(2)HPO(4) and preoxidation could significantly affect the physicochemical properties of prepared activated carbons. The apparent surface area and total pore volume as high as 1154 m(2)/g and 0.670 cm(3)/g were obtained respectively, when combined process of impregnation followed by preoxidation at 200°C and activation at 700°C was carried out. Meanwhile, the activated carbon yield and maximum methylene blue adsorption capacity up to 41.14% and 129.5 mg/g were achieved, respectively. The results exhibited that (NH(4))(2)HPO(4) could be an effective activating agent for producing activated carbons from rice straw.
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Pricing and availability is not currently available.
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Pricing and availability is not currently available.