Merck
Model-based design of peptide chromatographic purification processes.
Journal of chromatography. A (2013-03-08)
David Gétaz, Guido Stroehlein, Alessandro Butté, Massimo Morbidelli
ABSTRACT
In this work we present a general procedure for the model-based optimization of a polypeptide crude mixture purification process through its application to a case of industrial relevance. This is done to show how much modeling can be beneficial to optimize complex chromatographic processes in the industrial environment. The target peptide elution profile was modeled with a two sites adsorption equilibrium isotherm exhibiting two inflection points. The variation of the isotherm parameters with the modifier concentration was accounted for. The adsorption isotherm parameters of the target peptide were obtained by the inverse method. The elution of the impurities was approximated by lumping them into pseudo-impurities and by regressing their adsorption isotherm parameters directly as a function of the corresponding parameters of the target peptide. After model calibration and validation by comparison with suitable experimental data, Pareto optimizations of the process were carried out so as to select the optimal batch process.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)