Merck
  • Immobilization and stabilization of cholesterol oxidase on modified sepharose particles.
Immobilization and stabilization of cholesterol oxidase on modified sepharose particles.
International journal of biological macromolecules (2013-02-12)
Yi Chen, Yu Xin, Hailin Yang, Ling Zhang, Yuran Zhang, Xiaole Xia, Yanjun Tong, Wu Wang
ABSTRACT
For the cholesterol oxidase from Brevibacterium sp. M201008 was not stable as free enzyme form, it had been covalently immobilized onto functionalized sepharose particles that were activated with N-ethyl-N'-3-dimethylaminopropyl carbodiimide (EDC).The optimal conditions of enzyme immobilization were determined, and the immobilized enzyme activity was 18.03 U/g of support. The surface morphology and thermal behavior of the immobilized enzyme were observed by scanning electron microscopy and differential scanning calorimetry, respectively. The binding of the enzyme to support was confirmed using Fourier transform infrared spectroscopy. The results demonstrated that the thermal, pH, and storage stabilities of cholesterol oxidase were increased by immobilization. More than 90% of the initial activity of the immobilized enzyme was remained at the points of pH 4.0 and 9.0, and that was much more stable than it was free enzyme form. Under same storage conditions, the free enzyme lost 97.8% of its initial activity after 45 h, whereas the immobilized enzyme only lost its 35.8% activity.
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Agarose, Medium EEO, Molecular Biology
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Wide range, Molecular Biology
Sigma-Aldrich
Agarose, Molecular Biology
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO