Merck
  • A cocrystal of two Mo(VI) complexes bearing different diastereomers of the 2,4-di-tert-butyl-6-{[(1-oxido-1-phenylpropan-2-yl)(methyl)amino]methyl}phenolate ligand derived from (+)-ephedrine.
A cocrystal of two Mo(VI) complexes bearing different diastereomers of the 2,4-di-tert-butyl-6-{[(1-oxido-1-phenylpropan-2-yl)(methyl)amino]methyl}phenolate ligand derived from (+)-ephedrine.
Acta crystallographica. Section C, Crystal structure communications (2013-05-01)
Reijo Sillanpää, Mikko M Hänninen
ABSTRACT
The title cocrystal contains two chiral conformational diastereomers, viz. (1S,2R,RN)- and (1S,2R,SN)-, of [2,4-di-tert-butyl-6-{[(1-oxido-1-phenylpropan-2-yl)(methyl)amino]methyl}phenolato](methanol)-cis-dioxidomolybdenum(VI), [Mo(C25H35NO2)O2(CH3OH)], representing the first example of a structurally characterized molybdenum complex with enantiomerically pure ephedrine derivative ligands. The Mo(VI) cations exhibit differently distorted octahedral coordination environments, with two oxide ligands positioned cis to each other. The remainder of the coordination comprises phenoxide, alkoxide and methanol O atoms, with an amine N atom completing the octahedron. The distinct complexes are linked by strong intermolecular O-H···O hydrogen bonds, resulting in one-dimensional molecular chains. Furthermore, the phenyl rings are involved in weak T-shaped/edge-to-face π-π interactions with each other.
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis