Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Tensile characterization of porcine temporomandibular joint disc attachments.
Tensile characterization of porcine temporomandibular joint disc attachments.
Journal of dental research (2013-06-21)
M K Murphy, B Arzi, J C Hu, K A Athanasiou
ABSTRACT
The frequency and impact of temporomandibular joint (TMJ) disorders necessitate research in characterizing the joint's function. The 6 discal attachments have not yet been systematically characterized under tension. Understanding their role in joint function may guide our study of TMJ pathologies, including disc displacement. In the present study, a porcine model was used to characterize the attachments in tension anteroposteriorly and mediolaterally, based on previously identified similarities in the porcine and human masticatory behaviors and discal properties. Tensile stiffness, strength, toughness, and maximum strain were quantified. Collagen alignment was characterized via polarized light and scanning electron microscopy. Anisotropy was demonstrated in all attachments, with the exception of the anterior inferior attachment. Anteroposteriorly, the lateral attachment was stiffest (8.3 MPa) and the anterior superior was least stiff (1.4 MPa). Mediolaterally, the posterior superior attachment was stiffest (16.3 MPa) and the medial was least stiff (1.4 MPa). The greatest strain was observed in the lateral attachment in the mediolateral direction and the posterior superior attachment in the anteroposterior direction. With greatest strains in the most commonly observed directions of disc displacement, it is suggested that compromise in the posterior and lateral attachments contributes to partial lateral and anterior disc displacement.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture