Merck
  • Mechanism for transforming cytosolic SOD1 into integral membrane proteins of organelles by ALS-causing mutations.
Mechanism for transforming cytosolic SOD1 into integral membrane proteins of organelles by ALS-causing mutations.
Biochimica et biophysica acta (2014-10-14)
Liangzhong Lim, Xiaowen Lee, Jianxing Song
ABSTRACT
Mutations in superoxide dismutase 1 (SOD1) cause familial amyotrophic lateral sclerosis (FALS), while wild-type SOD1 has been implicated in sporadic ALS (SALS). SOD1 mutants are now recognized to acquire one or more toxicities that include their association with mitochondrial and endoplasmic reticulum membranes but the underlying structural mechanism remains unknown. Here we determine NMR conformations of both wild-type and a truncation mutant (L126Z) of SOD1 in aqueous solution and a membrane environment. The truncation mutant (which causes FALS at very low levels, indicating its elevated toxicity) is highly unstructured in solution, failing to adopt the β-barrel SOD1 native structure. Wild-type SOD1 is also highly unstructured upon reduction of disulfides and depletion of zinc. Most remarkably, both mutant and wild type adopt similar, highly-helical conformations in a membrane environment. Thus, either truncation or depletion of zinc is sufficient to eliminate the native β-barrel structure, and transform cytosolic SOD1 into membrane proteins energetically driven by forming amphiphilic helices in membranes. That zinc-deficiency is sufficient to produce a similar transformation in wild-type SOD1 implies that the wild-type and FALS-linked SOD1 mutants may trigger ALS by a common mechanism.
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, wire reel, 100m, diameter 0.25mm, as drawn, 99.99+%
Zinc, foil, 4mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%