Merck
  • Exploring the potential association between brominated diphenyl ethers, polychlorinated biphenyls, organochlorine pesticides, perfluorinated compounds, phthalates, and bisphenol A in polycystic ovary syndrome: a case-control study.
Exploring the potential association between brominated diphenyl ethers, polychlorinated biphenyls, organochlorine pesticides, perfluorinated compounds, phthalates, and bisphenol A in polycystic ovary syndrome: a case-control study.
BMC endocrine disorders (2014-10-29)
Sara J Vagi, Eduardo Azziz-Baumgartner, Andreas Sjödin, Antonia M Calafat, Daniel Dumesic, Leonardo Gonzalez, Kayoko Kato, Manori J Silva, Xiaoyun Ye, Ricardo Azziz
ABSTRACT
Polycystic Ovary Syndrome (PCOS) is an endocrine-metabolic disorder that affects approximately 6-10% of women of child-bearing age. Although preliminary studies suggest that certain pollutants may act as endocrine disruptors in animals, little is known about their potential association with PCOS. The objective of this case-control pilot study is to determine whether women with PCOS have higher concentrations of specific environmental contaminants compared to women who have not developed PCOS. Fifty-two PCOS case-patients (diagnosed using the National Institutes of Health 1990 definition) and 50 controls were recruited in 2007-2008, from an urban academic medical center in Los Angeles, CA. Brominated diphenyl ethers, polychlorinated biphenyls (PCBs), organochlorine pesticides, and perfluorinated compounds (PFCs) were measured in serum, and phthalates metabolites and bisphenol A (BPA) in urine. PCOS case-patients had significantly higher geometric mean (GM) serum concentrations of two PFCs: perfluorooctanoate (PFOA) (GMcases = 4.1 μg/L, GMcontrols = 2.3 μg/L; p = 0.001) and perfluorooctane sulfonate (PFOS) (GMcases = 8.2 μg/L, GMcontrols = 4.9 μg/L; p = 0.01), and lower urinary concentrations of monobenzyl phthalate (mBzP) (GMcases = 7.5 μg/g creatinine, GMcontrols = 11.7 μg/g creatinine; p = 0.02). Logistic regression, controlling for body mass index, age and race, identified an increased likelihood of PCOS in subjects with higher serum concentrations of PFOA and PFOS (adjusted-ORs = 5.8-6.9, p < 0.05), and with lower urine concentrations of mBzP and mono-n-butyl phthalate (mBP) (aORs = 0.14-0.25, p < 0.05). Our data suggest that PCOS case-patients may differ from controls in their environmental contaminant profile. PCOS subjects had higher serum concentrations of two PFCs, PFOA and PFOS, and lower urine concentrations of mBP and mBzP. Future studies are needed to confirm these preliminary findings and determine if these chemicals or their precursors may have a role in the pathogenesis of PCOS.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Methanol, analytical standard
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material