Merck
  • Effects of naltrexone plus topiramate on ethanol self-administration and tyrosine hydroxylase gene expression changes.
Effects of naltrexone plus topiramate on ethanol self-administration and tyrosine hydroxylase gene expression changes.
Addiction biology (2013-04-12)
Francisco Navarrete, Gabriel Rubio, Jorge Manzanares
ABSTRACT
The purpose of this study was to evaluate the effects of naltrexone (0.7 mg/kg) and/or topiramate (25 mg/kg) on ethanol consumption and the motivation to drink in an oral-operant conditioning paradigm in C57BL/6 mice. Subsequent real-time polymerase chain reaction (PCR) experiments were performed to analyze gene expression changes in tyrosine hydroxylase (TH) in the ventral tegmental area (VTA). The administration of naltrexone significantly reduced ethanol consumption and the motivation to drink during the different stages of the experiment, whereas the treatment with topiramate resulted in a much lower effect. Interestingly, the administration of naltrexone plus topiramate reduced ethanol consumption markedly compared with single-drug treatment. The water self-administration paradigm was also performed using the same drugs and no differences were found between treatment groups. Real-time PCR analyses revealed that naltrexone significantly normalized the increase of TH gene expression in the VTA induced by ethanol, whereas the administration of topiramate did not produce any significant effect. In the ethanol self-administration procedure, the combination of both drugs further reduced TH gene expression, reaching statistical significance compared with the vehicle, naltrexone or topiramate groups. Taken together, these findings indicate that the administration of naltrexone plus topiramate further reduced ethanol consumption and the motivation to drink in comparison with single-drug treatment. This action may be due, at least in part, to a greater decrease in TH gene expression in the VTA. These results suggest that the combination of both drugs deserves further exploration for the treatment of problems related to alcohol consumption.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology