Merck
  • Hot-melts in buccoadhesive patches: an approach for bioavailability enhancement of highly-metabolized drugs with short elimination half-life.
Hot-melts in buccoadhesive patches: an approach for bioavailability enhancement of highly-metabolized drugs with short elimination half-life.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
Galal M El Mahrouk, Omaima N ElGazayerly, Ahmed A Aboelwafa, Maie S Taha
ABSTRACT
The present study deals with the inclusion or incorporation of hot-melts into buccoadhesive patches. Our aim is to develop a patient-friendly dosage form that is capable of extending release of short elimination half-life drugs so to decrease dosing frequency and to increase the bioavailability of highly-metabolized drugs with the ultimate aim of dose reduction. Tizanidine hydrochloride (TIZ) was used as a model drug.TIZ was incorporated into Compritol-based hot-melts, and then further formulated into buccal patches prepared using HPMC, PVA and Polyox. A Central Composite Face-centered Design was employed to statistically optimize the formulation variables; HPMC solution/PVA solution weight ratio, Compritol/TIZ ratio in the hot-melts and percentage Polyox. The optimized formula suggested by the software was successful in controlling drug release, where 85% of TIZ was released after 4 h and the patch showed acceptable mucoadhesion properties. Pharmacokinetic parameters of TIZ from the optimized formula were compared to those of the immediate release tablet, Sirdalud®, as reference in human volunteers using a randomized crossover design. Significant increase was observed for Cmax, Tmax, AUC(0-12) and AUC(0-1). The increase in relative bioavailability of TIZ from the optimized formula was 2.57 folds.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard