Merck
  • Intratumoral heterogeneity of ADAM23 promotes tumor growth and metastasis through LGI4 and nitric oxide signals.
Intratumoral heterogeneity of ADAM23 promotes tumor growth and metastasis through LGI4 and nitric oxide signals.
Oncogene (2014-03-26)
E T Costa, G F Barnabé, M Li, A A M Dias, T R Machado, P F Asprino, F P Cavalher, E N Ferreira, M Del Mar Inda, M H Nagai, B Malnic, M L Duarte, K R M Leite, A C S D de Barros, D M Carraro, R Chammas, H A Armelin, W Cavenee, F Furnari, A A Camargo
ABSTRACT
Intratumoral heterogeneity (ITH) represents an obstacle for cancer diagnosis and treatment, but little is known about its functional role in cancer progression. The A Desintegrin And Metalloproteinase 23 (ADAM23) gene is epigenetically silenced in different types of tumors, and silencing is often associated with advanced disease and metastasis. Here, we show that invasive breast tumors exhibit significant ADAM23-ITH and that this heterogeneity is critical for tumor growth and metastasis. We demonstrate that while loss of ADAM23 expression enhances invasion, it causes a severe proliferative deficiency and is not itself sufficient to trigger metastasis. Rather, we observed that, in ADAM23-heterotypic environments, ADAM23-negative cells promote tumor growth and metastasis by enhancing the proliferation and invasion of adjacent A23-positive cells through the production of LGI4 (Leucine-rich Glioma Inactivated 4) and nitric oxide (NO). Ablation of LGI4 and NO in A23-negative cells significantly attenuates A23-positive cell proliferation and invasion. Our work denotes a driving role of ADAM23-ITH during disease progression, shifting the malignant phenotype from the cellular to the tissue level. Our findings also provide insights for therapeutic intervention, enforcing the need to ascertain ITH to improve cancer diagnosis and therapy.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard