Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Sensitive and selective determination of hydroxychloroquine in the presence of uric acid using a new nanostructure self-assembled monolayer modified electrode: optimization by multivariate data analysis.
Sensitive and selective determination of hydroxychloroquine in the presence of uric acid using a new nanostructure self-assembled monolayer modified electrode: optimization by multivariate data analysis.
The Analyst (2014-06-24)
Asma Khoobi, Sayed Mehdi Ghoreishi, Mohsen Behpour
ABSTRACT
A highly sensitive electrochemical nanosensor was developed using covalent modification of a glassy carbon electrode (GCE) by self-assembly of a novel Schiff base. Scanning electron microscopy (SEM) and electrochemical techniques were used to investigate the immobilization of the self-assembled monolayer (SAM) on the GCE. The electrochemical behavior of hydroxychloroquine (HCQ) in the presence of uric acid (UA) at the surface of the modified electrode was studied using the differential pulse voltammetry (DPV) technique. Response surface methodology (RSM) is used to optimize the effects of various operating variables such as pH, immersion time, scan rate, step potential and modulation amplitude on the voltammetric response of HCQ. RSM formulates a mathematical model which correlates the independent parameters with the peak current of HCQ. The central composite rotatable design (CCRD) has been applied to conduct the experiments. Then, under the optimized conditions, HCQ was determined in the presence of UA. The electrochemical measurements demonstrated that this biosensor responded well to HCQ, confirming that the self-assembly immobilization method was effective. Also, the interference, the storage stability, and the reproducibility of the biosensor were studied and assessed. The developed nanosensor was economical and efficient, making it potentially attractive for application to real sample analysis.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B