Merck
  • Investigation on cellular interactions of astrocytes with zinc oxide nanoparticles using rat C6 cell lines.
Investigation on cellular interactions of astrocytes with zinc oxide nanoparticles using rat C6 cell lines.
Colloids and surfaces. B, Biointerfaces (2015-06-10)
S Sruthi, P V Mohanan
ABSTRACT
Zinc oxide nanoparticles (ZnO NPs) are widely used in cosmetic industries and have also found important applications in electrical and chemical industries. It is well documented that inhaled ZnO NPs can reach the brain through the olfactory neuronal pathway and can interfere with the brain zinc homeostasis. Most of the studies focus on the toxicity of ZnO NPs on neuronal cells and microglia. Not much work is available on the bio interaction of ZnO NPs with astrocytes, the major cells involved in brain homeostasis. Therefore, this study focuses on the interaction of ZnO NPs with rat C6 glial cells. The results of this study reveal that the nanoparticles are taken up by the astrocytes and induce a time and dose dependent toxicological response which is indicated by the nanoparticle uptake studies and cell viability assays. Also the results of DCFH-DA (2',7'-dichlorofluorescein diacetate) assay show that the ZnO NPs induce strong oxidative stress in cells at 3 and 6h. However, at 24h the reactive oxygen species (ROS) detected in the nanoparticles treated groups were same as that of control. The mode of cell death induced by ZnO NPs was apoptosis as revealed by the nuclear condensation studies, live dead assay using acridine orange/ethidium bromide and apoptosis detection kit. This study, which explores the interaction of ZnO NPs with astrocytes, concludes that the persistence of particle can continue to have a damaging effect on the astrocytes. And hence the time of exposure and particle clearance by the system should be evaluated more thoroughly to prevent the health hazards due to these particles.
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder