Merck
  • Coexistence of and interaction relationships between an aflatoxin-producing fungus and a bacterium.
Coexistence of and interaction relationships between an aflatoxin-producing fungus and a bacterium.
Fungal biology (2015-06-11)
Quan-Hong Yan, Jian-Xiang Zhou, Hong-Zhou Li, Qing-Qing Zhi, Xiao-Ping Zhou, Zhu-Mei He
ABSTRACT
The interactions between aflatoxin-producing fungi and bacteria have opened up a new avenue for identifying biological agents suitable for controlling aflatoxin contamination. In this study, we analysed the interactions between A. flavus and the bacterium Burkholderia gladioli M3 that coexist in rice that is naturally contaminated with A. flavus. Our results showed that a cell-free culture filtrate (CCF) and the metabolite bongkrekic acid of the M3 strain potently suppressed the mycelial growth and spore production, and then affected the production of aflatoxin of A. flavus. Bongkrekic acid secreted by the M3 strain exhibited higher antifungal activity than did analogues. The CCF of the M3 strain and its metabolite bongkrekic acid can inhibit the growth of A. flavus, but the metabolites of A. flavus, aflatoxins, exerted no inhibitory effect on the growth of the M3 strain. Furthermore, we determined that the M3 cells could use the dead mycelia of A. flavus as energy sources for reproduction, while A. flavus could not grow in a solution containing dead M3 cells. In summary, these results indicated that B. gladioli has a competitive advantage in survival when it coexists with its fungal partner A. flavus.
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Ethyl acetate