Merck
  • Calcium/calmodulin-dependent protein kinase regulates the PINK1/Parkin and DJ-1 pathways of mitophagy during sepsis.
Calcium/calmodulin-dependent protein kinase regulates the PINK1/Parkin and DJ-1 pathways of mitophagy during sepsis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-06-16)
Xianghong Zhang, Du Yuan, Qian Sun, Li Xu, Emma Lee, Anthony J Lewis, Brian S Zuckerbraun, Matthew R Rosengart
ABSTRACT
During sepsis and shock states, mitochondrial dysfunction occurs. Consequently, adaptive mechanisms, such as fission, fusion, and mitophagy, are induced to eliminate damaged portions or entire dysfunctional mitochondria. The regulatory PINK1/Parkin and DJ-1 pathways are strongly induced by mitochondrial depolarization, although a direct link between loss of mitochondrial membrane potential (Δ
Sigma-Aldrich
MISSION® esiRNA, targeting human KCNA1
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder