Skip to Content
Merck

BDPH9BCCDB05

Bide Pharmatech

5-(4-Methoxyphenyl)pentanoic acid

Synonym(s):

5-(4-methoxyphenyl)pentanoic acid

Select a Size

Change View

About This Item

Linear Formula:
C12H16O3
CAS Number:
Molecular Weight:
208.26
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H16O3/c1-15-11-8-6-10(7-9-11)4-2-3-5-12(13)14/h6-9H,2-5H2,1H3,(H,13,14)

Inchi Key

BGPOHJSGSYVQSU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 5-(4-Methoxyphenyl)pentanoic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library