Skip to Content
Merck

BDPH9BCD3BE5

Bide Pharmatech

1-Isopropyl-3-methyl-1H-pyrazole-5-carboxylic acid

Synonym(s):

1-isopropyl-3-methyl-1H-pyrazole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H12O2N2
Molecular Weight:
168.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H12N2O2/c1-5(2)10-7(8(11)12)4-6(3)9-10/h4-5H,1-3H3,(H,11,12)

Inchi Key

AUZHMOQAPWTGTB-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library