Skip to Content
Merck

BDPH9BCD3F2D

Bide Pharmatech

3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine

Synonym(s):

3-cyclopropyl-1-methyl-1H-pyrazol-5-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H11N3
CAS Number:
Molecular Weight:
137.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H11N3/c1-10-7(8)4-6(9-10)5-2-3-5/h4-5H,2-3,8H2,1H3

Inchi Key

YWHWPIRLFHZSFS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library