Skip to Content

BDPH9BCD4E76

Bide Pharmatech

2-Amino-6-bromoquinazolin-4-ol

Synonym(s):

2-amino-6-bromo-4-quinazolinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6O1N3Br1
CAS Number:
Molecular Weight:
240.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrN3O/c9-4-1-2-6-5(3-4)7(13)12-8(10)11-6/h1-3H,(H3,10,11,12,13)

Inchi Key

BDPCBRSHZVHPKX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Off white to pale-yellow solid



Still not finding the right product?

Explore all of our products under 2-Amino-6-bromoquinazolin-4-ol


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library