Skip to Content

BDPH9BCD7FF2

Bide Pharmatech

(2-Methyl-1H-benzo[d]imidazol-6-yl)boronic acid

Synonym(s):

(2-methyl-1H-benzo[d]imidazol-6-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O2N2B1
CAS Number:
Molecular Weight:
175.98
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9BN2O2/c1-5-10-7-3-2-6(9(12)13)4-8(7)11-5/h2-4,12-13H,1H3,(H,10,11)

Inchi Key

FZIFUKQWKWRSER-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library