Skip to Content
Merck

BDPH9BCD84FF

Bide Pharmatech

tert-Butyl (4-(aminomethyl)benzyl)(methyl)carbamate

Synonym(s):

tert-butyl 4-(aminomethyl)benzyl(methyl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H22O2N2
CAS Number:
Molecular Weight:
250.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H22N2O2/c1-14(2,3)18-13(17)16(4)10-12-7-5-11(9-15)6-8-12/h5-8H,9-10,15H2,1-4H3

Inchi Key

FXWZLSMMROZOGP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library