Skip to Content
Merck

BDPH9BCDECB3

Bide Pharmatech

Ethyl 1-((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

Synonym(s):

ethyl 1-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-2-ethoxy-1H-benzimidazole-7-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C26H23O3N3
CAS Number:
Molecular Weight:
425.49
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C26H23N3O3/c1-3-31-25(30)22-10-7-11-23-24(22)29(26(28-23)32-4-2)17-18-12-14-19(15-13-18)21-9-6-5-8-20(21)16-27/h5-15H,3-4,17H2,1-2H3

Inchi Key

GYTTUVXCCHRIBA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Off White to Pale-yellow to Yellow-brown Solid




hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library