Skip to Content
Merck

BDPH9C01EB87

Bide Pharmatech

O-(4-Iodobenzyl)hydroxylamine hydrochloride

Synonym(s):

O-(4-iodobenzyl)hydroxylamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O1N1Cl1I1
CAS Number:
Molecular Weight:
285.51
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8INO.ClH/c8-7-3-1-6(2-4-7)5-10-9;/h1-4H,5,9H2;1H

Inchi Key

QKTGWFUBZHRJRK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library