Skip to Content
Merck

Skip To

BL3H160C19B2

BLD Pharmatech

4-Chloro-2-(hydroxymethyl)phenol

Synonym(s):

4-chloro-2-(hydroxymethyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Ships in 5 days - from Aldrich Partner.
€46.00
5 g
Ships in 5 days - from Aldrich Partner.
€71.00
10 g
Ships in 5 days - from Aldrich Partner.
€112.00
25 g
Ships in 5 days - from Aldrich Partner.
€231.00
100 g
Ships in 10 days - from Aldrich Partner.
€651.00

About This Item

Linear Formula:
C7H7O2Cl1
CAS Number:
Molecular Weight:
158.58
MDL number:

Vendor SKU: BD4781-1G

€46.00


Ships in 5 days - from Aldrich Partner.

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7ClO2/c8-6-1-2-7(10)5(3-6)4-9/h1-3,9-10H,4H2

Inchi Key

AFPSETMCMFRVPG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to Pale-yellow Powder or Crystals




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library