Merck
HomeProductsLabwareLabware PartnersExcel Scientific Products
Excel Scientific Products
Photograph showing two gloved hands using an adhesive seal on a roller to seal a microplate.
Excel Scientific is a global supplier of microplate sealing films and foils engineered for applications including PCR, qPCR, ELISA, cell culture, cold and long-term storage, autosampling, and more. They offer a full product range to meet the specific needs of research and testing laboratories as well as biotech and pharmaceutical companies. Their materials, adhesive formulations, and design characteristics are custom-tailored to maximize performance.
  • SealPlate® Rollmate™ films in easy-to-roll formats
  • SealPlate® precut microplate seals with tabs for easy handling
  • Ministrips™ for sealing single or double microplate rows
  • ColorTabs™ for sample identification and labeling
  • ThinSeal™ for pierceability with single-channel pipet tips, probes, and needles
  • AeraSeal™, designed to facilitate air and CO2 exchange for cell culture applications
  • eXTReme™ Seal with extra strong adhesive to minimize evaporation without residue
Products
Showing 1-20 of 70
Change View
Showing 1-20 of 70
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 70 results