Merck
  • Effect of levonorgestrel IUD and oral medroxyprogesterone acetate on glandular and stromal progesterone receptors (PRA and PRB), and estrogen receptors (ER-alpha and ER-beta) in human endometrial hyperplasia.
Effect of levonorgestrel IUD and oral medroxyprogesterone acetate on glandular and stromal progesterone receptors (PRA and PRB), and estrogen receptors (ER-alpha and ER-beta) in human endometrial hyperplasia.
Gynecologic oncology (2005-12-06)
Anne Beate Vereide, Turid Kaino, Georg Sager, Marit Arnes, Anne Ørbo
ABSTRACT
The effect on progesterone and estrogen receptor expression in glands and stroma after two different treatment regimens of endometrial hyperplasia was determined. Pre- and post-treatment paraffin-embedded endometrial hyperplasia specimens from women treated with levonorgestrel (LNG) intrauterine device (n = 21) and women treated with 10 mg medroxyprogesterone acetate (MPA) for 10 days per cycle (n = 29) were examined immunohistochemically and evaluated by H-score (a semi-quantitative microscopical method evaluating staining intensity and number of stained cells, scale 0-3) for changes in expression of PRA (progesterone receptor A), PRB (progesterone receptor B), ER-alpha (estrogen receptor-alpha), ER-beta (estrogen receptor-beta) and AR (androgen receptors) after 3 months of treatment. All the patients in the LNG IUD group responded to treatment with no sign of hyperplasia after 3 months, while only about half of the patients given MPA orally responded. Expression of PRA, PRB, ER-alpha and ER-beta were markedly reduced after progestin treatment in both treatment groups but the reduction was much more pronounced in the LNG group (H-score for PRA was reduced from 2.61 to 0.11 in glands and from 2.26 to 0.09 in stroma in LNG group. Corresponding reduction for PRB in the LNG group was from 1.96 to 0.11 and from 0.83 to 0.01. PRA was reduced from 2.53 to 1.78 in glands and from 1.93 to 1.30 in stroma in the MPA group. Corresponding reduction for PRB in the MPA group was from 2.02 to 1.25 in glands and from 0.80 to 0.34 in stroma). Weak and focal stromal expression of AR was demonstrated in 22% of the specimens before but not after therapy. There was a statistically significant reduction in both PR and ER among the responders whereas non-responders showed no statistical change after treatment. The present study shows that LNG IUD causes an almost complete down-regulation (lack of immunohistochemical expression) of PR expression and a considerable down-regulation of ER expression in both glands and stroma. The changes in receptor expression were markedly less pronounced after treatment with intermittent oral MPA. The differences in receptor expression among responders and non-responders might serve as possible markers for therapy response.
Product Description
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)