Merck
  • Co-translational protein targeting facilitates centrosomal recruitment of PCNT during centrosome maturation in vertebrates.
Co-translational protein targeting facilitates centrosomal recruitment of PCNT during centrosome maturation in vertebrates.
eLife (2018-05-01)
Guadalupe Sepulveda, Mark Antkowiak, Ingrid Brust-Mascher, Karan Mahe, Tingyoung Ou, Noemi M Castro, Lana N Christensen, Lee Cheung, Xueer Jiang, Daniel Yoon, Bo Huang, Li-En Jao
ABSTRACT
As microtubule-organizing centers of animal cells, centrosomes guide the formation of the bipolar spindle that segregates chromosomes during mitosis. At mitosis onset, centrosomes maximize microtubule-organizing activity by rapidly expanding the pericentriolar material (PCM). This process is in part driven by the large PCM protein pericentrin (PCNT), as its level increases at the PCM and helps recruit additional PCM components. However, the mechanism underlying the timely centrosomal enrichment of PCNT remains unclear. Here, we show that PCNT is delivered co-translationally to centrosomes during early mitosis by cytoplasmic dynein, as evidenced by centrosomal enrichment of PCNT mRNA, its translation near centrosomes, and requirement of intact polysomes for PCNT mRNA localization. Additionally, the microtubule minus-end regulator, ASPM, is also targeted co-translationally to mitotic spindle poles. Together, these findings suggest that co-translational targeting of cytoplasmic proteins to specific subcellular destinations may be a generalized protein targeting mechanism.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Millipore
Emetine, Dihydrochloride, Principal alkaloid of ipecac, isolated from the ground roots of Uragoga ipecacuanha.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Cytoplasmic Dynein Inhibitor, Ciliobrevin D, Ciliobrevin D is a cell-permeable, reversible, and specific blocker of AAA+ ATPase motor cytoplasmic dynein. Disrupts spindle pole focusing and kinetochore-microtubule attachment (~10 to 40 µM).
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep