- Tacrine(10)-hupyridone, a dual-binding acetylcholinesterase inhibitor, potently attenuates scopolamine-induced impairments of cognition in mice.
Tacrine(10)-hupyridone, a dual-binding acetylcholinesterase inhibitor, potently attenuates scopolamine-induced impairments of cognition in mice.
Tacrine(10)-hupyridone (A10E) was designed as a dual-binding acetylcholinesterase (AChE) inhibitor from the modification of tacrine and a fragment of huperzine A. We have found that A10E effectively inhibited AChE in a mixed competitive manner, with an IC50 of 26.4 nM, which is more potent than those of tacrine and huperzine A. Most importantly, we have shown, for the first time that A10E attenuated scopolamine-induced cognitive impairments without affecting motor function in mice. A10E effectively attenuated impairments of learning and memory to a similar extent as donepezil, an inhibitor of AChE used for treating Alzheimer's disease (AD). In addition, A10E significantly decreased AChE activity in the brain of mice, suggesting that A10E might cross the brain blood-barrier. Taken together, our results demonstrated that A10E, a designed dual-binding AChE inhibitor, could effectively reverse cognitive impairments, indicating that A10E might provide therapeutic efficacy for AD treatment.