Merck
  • Hippocampal neurons require a large pool of glutathione to sustain dendrite integrity and cognitive function.
Hippocampal neurons require a large pool of glutathione to sustain dendrite integrity and cognitive function.
Redox biology (2018-08-15)
Seila Fernandez-Fernandez, Veronica Bobo-Jimenez, Raquel Requejo-Aguilar, Silvia Gonzalez-Fernandez, Monica Resch, Monica Carabias-Carrasco, Joaquim Ros, Angeles Almeida, Juan P Bolaños
ABSTRACT
Loss of brain glutathione has been associated with cognitive decline and neuronal death during aging and neurodegenerative diseases. However, whether decreased glutathione precedes or follows neuronal dysfunction has not been unambiguously elucidated. Previous attempts to address this issue were approached by fully eliminating glutathione, a strategy causing abrupt lethality or premature neuronal death that led to multiple interpretations. To overcome this drawback, here we aimed to moderately decrease glutathione content by genetically knocking down the rate-limiting enzyme of glutathione biosynthesis in mouse neurons in vivo. Biochemical and morphological analyses of the brain revealed a modest glutathione decrease and redox stress throughout the hippocampus, although neuronal dendrite disruption and glial activation was confined to the hippocampal CA1 layer. Furthermore, the behavioral characterization exhibited signs consistent with cognitive impairment. These results indicate that the hippocampal neurons require a large pool of glutathione to sustain dendrite integrity and cognitive function.
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture