Merck
  • Isolation and characterization of cytoplasmic poly(A) polymerase from cryptobiotic gastrulae of Artemia salina.
Isolation and characterization of cytoplasmic poly(A) polymerase from cryptobiotic gastrulae of Artemia salina.
European journal of biochemistry (1985-03-01)
E Roggen, H Slegers
ABSTRACT
Poly(A) polymerase has been purified to near homogeneity from the cytoplasm of Artemia salina cryptobiotic gastrulae by ion-exchange chromatography on DEAE-cellulose, DEAE-Sepharose CL-6B and phosphocellulose P11, gel filtration on CL-Sepharose 6B, affinity chromatography on poly(A)-Sepharose 4B and ATP-agarose. The enzyme is fully dependent on exogeneous oligo(riboadenylic acid) and is free of any nuclease or other enzyme activities. In standard assay conditions the enzyme preparation has a specific activity of 5.6 mumol AMP . h-1 . (mg protein)-1. Sodium dodecyl sulphate/polyacrylamide gel electrophoresis reveals the presence of only two proteins with Mr 94 000 and 70 000. The Mr-70 000 protein has been identified as poly(A) polymerase. The enzyme is exclusively activated by Mn2+. Addition of Ca2+, Mg2+, Zn2+, NH4+, K+ or Na+ inhibits the enzymatic reaction. The activity is specific for ATP and competitive inhibition is observed in the presence of other ribonucleoside 5'-triphosphates. AMP incorporation is time-dependent and is increased non-linearly with protein and primer concentration.
Sigma-Aldrich
Adenosine 5′-triphosphate, immobilized on Agarose 4B, suitable for affinity chromatography, powder (lyophilized)