Merck
  • Tandem-type Pd(II)-catalyzed oxidative Heck reaction/intramolecular C-H amidation sequence: a novel route to 4-aryl-2-quinolinones.
Tandem-type Pd(II)-catalyzed oxidative Heck reaction/intramolecular C-H amidation sequence: a novel route to 4-aryl-2-quinolinones.
Chemical communications (Cambridge, England) (2012-03-24)
Kiyofumi Inamoto, Junpei Kawasaki, Kou Hiroya, Yoshinori Kondo, Takayuki Doi
ABSTRACT
A novel catalytic method for synthesizing 4-aryl-2-quinolinones is reported. The process involves two mechanistically independent, sequential Pd(II)-catalyzed reactions--the oxidative Heck reaction and the intramolecular C-H amidation--both of which smoothly proceed in the presence of a single catalytic system in a one-pot manner.
Sigma-Aldrich
4-Methoxycarbonylphenylboronic acid, ≥95%
Sigma-Aldrich
p-Tolylboronic acid, 97%
Sigma-Aldrich
4-Fluorophenylboronic acid, ≥95%
Sigma-Aldrich
4-Methoxyphenylboronic acid, ≥95.0%
Sigma-Aldrich
4-Chlorophenylboronic acid, 95%
Sigma-Aldrich
4-(Trifluoromethyl)phenylboronic acid, ≥95.0%
Sigma-Aldrich
4-Bromophenylboronic acid, ≥95.0%