Merck
  • Synthesis and structure-activity relationship studies of 1,3-diarylprop-2-yn-1-ones: dual inhibitors of cyclooxygenases and lipoxygenases.
Synthesis and structure-activity relationship studies of 1,3-diarylprop-2-yn-1-ones: dual inhibitors of cyclooxygenases and lipoxygenases.
Journal of medicinal chemistry (2006-03-03)
P N Praveen Rao, Qiao-Hong Chen, Edward E Knaus
ABSTRACT
A group of 1,3-diarylprop-2-yn-1-ones (13, 17, 23, 26 and 27) possessing a C-3 p-SO2Me COX-2 pharmacophore were designed, synthesized and evaluated as potential dual inhibitors of cyclooxygenase-1/2 (COX-1/2) and 5/15-lipoxygenases (5/15-LOX) that exhibit vivo antiinflammatory and analgesic activities. Among this class of compounds, 3-(4-methanesulfonylphenyl)-1-(4-fluorophenyl)prop-2-yn-1-one (13h) was identified as a potent and selective inhibitor of COX-2 (COX-2 IC50 = 0.1 microM; SI = 300), being 5-fold more potent than rofecoxib (COX-2 IC50 = 0.5 microM; SI > 200). In a rat carrageenan-induced paw edema assay 13h exhibited moderate antiinflammatory activity (26% inhibition of inflammation) at 3 h after administration of a 30 mg/kg oral dose. A related dual COX-1/2 and 5/15-LOX inhibitor 3-(4-methanesulfonylphenyl)-1-(4-cyanophenyl)prop-2-yn-1-one (13g, COX-1 IC50 = 31.5 microM; COX-2 IC50 = 1.0 microM; SI = 31.5; 5-LOX IC50 = 1.0 microM; 15-LOX IC50 = 3.2 microM) exhibited more potent antiinflammatory activity (ED50 = 90 mg/kg), being superior to the reference drug aspirin (ED50 = 129 mg/kg). Within this group of compounds 3-(4-methanesulfonylphenyl)-1-(4-isopropylphenyl)prop-2-yn-1-one (13e) emerged as having an optimal combination of in vitro COX-1/2 and 5/15-LOX inhibitory effects (COX-1 IC50 = 9.2 microM; COX-2 IC50 = 0.32 microM; SI = 28; 5-LOX IC50 = 0.32 microM; 15-LOX IC50 = 0.36 microM) in conjunction with a good antiinflammatory activity (ED50 = 35 mg/kg) compared to the reference drug celecoxib (ED50 = 10.8 mg/kg) when administered orally. A molecular modeling study where 13e was docked in the COX-2 binding site indicated the C-1 p-i-Pr group was positioned within a hydrophobic pocket (Phe205, Val344, Val349, Phe381 and Leu534), and that this positioning of the i-Pr group facilitated orientation of the C-3 p-SO2Me COX-2 pharmacophore such that it inserted into the COX-2 secondary pocket (His90, Arg513, Ile517 and Val523). A related docking study of 13e in the 15-LOX binding site indicates that the C-3 p-SO2Me COX-2 pharmacophore was positioned in a region closer to the catalytic iron site where it undergoes a hydrogen bonding interaction with His541 and His366, and that the C-1 p-i-Pr substituent is buried deep in a hydrophobic pocket (Ile414, Ile418, Met419 and Ile593) near the base of the 15-LOX binding site.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard