Merck
  • Small molecule obatoclax (GX15-070) antagonizes MCL-1 and overcomes MCL-1-mediated resistance to apoptosis.
Small molecule obatoclax (GX15-070) antagonizes MCL-1 and overcomes MCL-1-mediated resistance to apoptosis.
Proceedings of the National Academy of Sciences of the United States of America (2007-11-28)
Mai Nguyen, Richard C Marcellus, Anne Roulston, Mark Watson, Lucile Serfass, S R Murthy Madiraju, Daniel Goulet, Jean Viallet, Laurent Bélec, Xavier Billot, Stephane Acoca, Enrico Purisima, Adrian Wiegmans, Leonie Cluse, Ricky W Johnstone, Pierre Beauparlant, Gordon C Shore
ABSTRACT
Elevated expression of members of the BCL-2 pro-survival family of proteins can confer resistance to apoptosis in cancer cells. Small molecule obatoclax (GX15-070), which is predicted to occupy a hydrophobic pocket within the BH3 binding groove of BCL-2, antagonizes these members and induces apoptosis, dependent on BAX and BAK. Reconstitution in yeast confirmed that obatoclax acts on the pathway and overcomes BCL-2-, BCL-XL-, BCL-w-, and MCL-1-mediated resistance to BAX or BAK. The compound potently interfered with the direct interaction between MCL-1 and BAK in intact mitochondrial outer membrane and inhibited the association between MCL-1 and BAK in intact cells. MCL-1 has been shown to confer resistance to the BCL-2/BCL-XL/BCL-w-selective antagonist ABT-737 and to the proteasome inhibitor bortezomib. In both cases, this resistance was overcome by obatoclax. These findings support a rational clinical development opportunity for the compound in cancer indications or treatments where MCL-1 contributes to resistance to cell killing.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Supelco
(−)-Epigallocatechin, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%