Merck
  • Rab7-harboring vesicles are carriers of the transferrin receptor through the biosynthetic secretory pathway.
Rab7-harboring vesicles are carriers of the transferrin receptor through the biosynthetic secretory pathway.
Science advances (2021-02-02)
Maika S Deffieu, Ieva Cesonyte, François Delalande, Gaelle Boncompain, Cristina Dorobantu, Eli Song, Vincent Lucansky, Aurélie Hirschler, Sarah Cianferani, Franck Perez, Christine Carapito, Raphael Gaudin
ABSTRACT
The biosynthetic secretory pathway is particularly challenging to investigate as it is underrepresented compared to the abundance of the other intracellular trafficking routes. Here, we combined the retention using selective hook (RUSH) to a CRISPR-Cas9 gene editing approach (eRUSH) and identified Rab7-harboring vesicles as an important intermediate compartment of the Golgi-to-plasma membrane transport of neosynthesized transferrin receptor (TfR). These vesicles did not exhibit degradative properties and were not associated to Rab6A-harboring vesicles. Rab7A was transiently associated to neosynthetic TfR-containing post-Golgi vesicles but dissociated before fusion with the plasma membrane. Together, our study reveals a role for Rab7 in the biosynthetic secretory pathway of the TfR, highlighting the diversity of the secretory vesicles' nature.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human PRICKLE1
Sigma-Aldrich
Triton X-100, laboratory grade