Merck
  • Architecture and organization of mouse posterior parietal cortex relative to extrastriate areas.
Architecture and organization of mouse posterior parietal cortex relative to extrastriate areas.
The European journal of neuroscience (2018-11-21)
Karoline Hovde, Michele Gianatti, Menno P Witter, Jonathan R Whitlock
ABSTRACT
The posterior parietal cortex (PPC) is a multifaceted region of cortex, contributing to several cognitive processes, including sensorimotor integration and spatial navigation. Although recent years have seen a considerable rise in the use of rodents, particularly mice, to investigate PPC and related networks, a coherent anatomical definition of PPC in the mouse is still lacking. To address this, we delineated the mouse PPC, using cyto- and chemoarchitectural markers from Nissl-, parvalbumin-and muscarinic acetylcholine receptor M2-staining. Additionally, we performed bilateral triple anterograde tracer injections in primary visual cortex (V1) and prepared flattened tangential sections from one hemisphere and coronal sections from the other, allowing us to co-register the cytoarchitectural features of PPC with V1 projections. This revealed that extrastriate area A was largely contained within lateral PPC, that medial PPC overlapped with the anterior portion of area AM, and that anterior RL overlapped partially with area PtP. Furthermore, triple anterograde tracer injections in PPC showed strong projections to associative thalamic nuclei as well as higher visual areas, orbitofrontal, cingulate and secondary motor cortices. Retrograde circuit mapping with rabies virus further showed that all cortical connections were reciprocal. These combined approaches provide a coherent definition of mouse PPC that incorporates laminar architecture, extrastriate projections, thalamic, and cortico-cortical connections.
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m2 Antibody, clone M2-2-B3, clone M2-2-B3, Chemicon®, from rat