Merck
  • Assembly and Cryo-EM structure determination of yeast mitochondrial RNA polymerase initiation complex intermediates.
Assembly and Cryo-EM structure determination of yeast mitochondrial RNA polymerase initiation complex intermediates.
STAR protocols (2021-04-20)
Sergio E Martinez, Anupam Singh, Brent De Wijngaert, Shemaila Sultana, Chhaya Dharia, Hans Vanbuel, Jiayu Shen, Daniel Vasilchuk, Smita S Patel, Kalyan Das
ABSTRACT
In yeast mitochondria, transcription initiation requires assembly of mitochondrial RNA polymerase and transcription initiation factor MTF1 at the DNA promoter initiation site. This protocol describes the purification of the component proteins and assembly of partially melted and fully melted initiation complex states. Both states co-exist in equilibrium in the same sample as seen by cryoelectron microscopy (cryo-EM) and allow elucidation of MTF1's structural roles in controlling the transition into elongation. We further outline how analysis of the complex by light scattering, thermal shift assay, and ultrafiltration assay exhibits reproducible results. For complete details on the use and execution of this protocol, please refer to De Wijngaert et al. (2021).
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture