Merck
TGFBR3L is an inhibin B co-receptor that regulates female fertility.
Science advances (2021-12-16)
Emilie Brûlé, Ying Wang, Yining Li, Yeu-Farn Lin, Xiang Zhou, Luisina Ongaro, Carlos A I Alonso, Evan R S Buddle, Alan L Schneyer, Chang-Hyeock Byeon, Cynthia S Hinck, Natalia Mendelev, John P Russell, Mitra Cowan, Ulrich Boehm, Frederique Ruf-Zamojski, Michel Zamojski, Cynthia L Andoniadou, Stuart C Sealfon, Craig A Harrison, Kelly L Walton, Andrew P Hinck, Daniel J Bernard
ABSTRACT
Follicle-stimulating hormone (FSH), a key regulator of ovarian function, is often used in infertility treatment. Gonadal inhibins suppress FSH synthesis by pituitary gonadotrope cells. The TGFβ type III receptor, betaglycan, is required for inhibin A suppression of FSH. The inhibin B co-receptor was previously unknown. Here, we report that the gonadotrope-restricted transmembrane protein, TGFBR3L, is the elusive inhibin B co-receptor. TGFBR3L binds inhibin B but not other TGFβ family ligands. TGFBR3L knockdown or overexpression abrogates or confers inhibin B activity in cells. Female Tgfbr3l knockout mice exhibit increased FSH levels, ovarian follicle development, and litter sizes. In contrast, female mice lacking both TGFBR3L and betaglycan are infertile. TGFBR3L’s function and cell-specific expression make it an attractive new target for the regulation of FSH and fertility.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid