Merck
  • IgE and IgG4 Epitope Mapping of Food Allergens with a Peptide Microarray Immunoassay.
IgE and IgG4 Epitope Mapping of Food Allergens with a Peptide Microarray Immunoassay.
Methods in molecular biology (Clifton, N.J.) (2022-09-25)
Javier Martínez-Botas, Carlos Fernández-Lozano, Aida Vaquero-Rey, Belén de la Hoz
ABSTRACT
Peptide microarrays are a powerful tool to identify linear epitopes of food allergens in a high-throughput manner. The main advantages of the microarray-based immunoassay are as follows: the possibility to assay thousands of targets simultaneously, the requirement of a low volume of serum, the more robust statistical analysis, and the possibility to test simultaneously several immunoglobulin subclasses. Among them, the last one has a special interest in the field of food allergy, because the development of tolerance to food allergens has been associated with a decrease in IgE and an increase in IgG4 levels against linear epitopes. However, the main limitation to the clinical use of microarray is the automated analysis of the data. Recent studies mapping the linear epitopes of food allergens with peptide microarray immunoassays have identified peptide biomarkers that can be used for early diagnosis of food allergies and to predict their severity or the self-development of tolerance. Using this approach, we have worked on epitope mapping of the two most important food allergens in the Spanish population, cow's milk, and chicken eggs. The final aim of these studies is to define subsets of peptides that could be used as biomarkers to improve the diagnosis and prognosis of food allergies. This chapter describes the protocol to produce microarrays using a library of overlapping peptides corresponding to the primary sequences of food allergens and data acquisition and analysis of IgE and IgG4 binding epitopes.
Sigma-Aldrich
α-Lactalbumin from bovine milk, Type III, calcium depleted, ≥85% (PAGE), lyophilized powder
Sigma-Aldrich
κ-Casein from bovine milk, ≥70% (PAGE), lyophilized powder
Sigma-Aldrich
α-Casein from bovine milk, ≥70% αs-casein basis (electrophoresis), lyophilized powder
Sigma-Aldrich
β-Casein from bovine milk, BioUltra, ≥98% (PAGE)
Sigma-Aldrich
Casein, Dephosphorylated from bovine milk, lyophilized powder
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder