Merck
  • Protocol to generate a patient derived xenograft model of acquired resistance to immunotherapy in humanized mice.
Protocol to generate a patient derived xenograft model of acquired resistance to immunotherapy in humanized mice.
STAR protocols (2022-11-02)
Alex Martínez-Sabadell, Pablo Ovejero Romero, Joaquín Arribas, Enrique J Arenas
ABSTRACT
Immunotherapy has revolutionized cancer treatment, but preclinical models are required to understand immunotherapy resistance mechanisms underlying patient relapse. This protocol describes how to generate an acquired resistance humanized in vivo model to immunotherapies in patient-derived xenografts (PDX). We detail steps to inject human CD34+ cells into NSG mice, followed by generation of immunoresistant PDX in humanized mice. This approach recapitulates the human immune system, allowing investigators to generate preclinical resistance models to different immunotherapies for identifying the resistant phenotype. For complete details on the use and execution of this protocol, please refer to Martínez-Sabadell et al., 2022 and Arenas et al. (2021).
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Millipore
EDTA, Disodium Salt, Dihydrate, Molecular Biology Grade
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein