Merck
  • Neutral antagonist activity of naltrexone and 6beta-naltrexol in naïve and opioid-dependent C6 cells expressing a mu-opioid receptor.
Neutral antagonist activity of naltrexone and 6beta-naltrexol in naïve and opioid-dependent C6 cells expressing a mu-opioid receptor.
British journal of pharmacology (2009-02-18)
M F Divin, F A Bradbury, F I Carroll, J R Traynor
ABSTRACT
Adenylyl cyclase sensitization occurs on chronic agonist activation of mu-opioid receptors and is manifested by an increase in cAMP levels (overshoot) on challenge with antagonist. It has been proposed that a long lasting constitutively active receptor is formed on chronic mu-opioid exposure and that antagonists with inverse agonist activity rapidly return the receptor to a basal state causing a cAMP overshoot and a more severe withdrawal response in vivo. This hypothesis depends on an accurate characterization of neutral and inverse agonist properties of opioid antagonists. C6 glioma and HEK293 cells expressing mu-opioid receptors were used. Opioid antagonists were examined for their ability to induce a cAMP overshoot following chronic treatment with the agonist DAMGO ([D-Ala(2),N-Me-Phe(4),Glyol(5)]-enkephalin). The compounds were also characterized as agonists, inverse agonists or neutral antagonists by using assays for competitive binding, [(35)S]GTPgammaS (guanosine-5'-O-(3-[(35)S]thio)triphosphate) binding and changes in cell surface receptor expression. Naltrexone, 6beta-naltrexol and naloxone were indistinguishable to the mu-opioid receptor in the opioid-naïve or dependent state and acted as neutral antagonists. The delta-opioid receptor inverse agonist RTI-5989-25 [(+)-N-[trans-4'-(2-methylphenyl)-2'-butenyl]-(3R,4R)-dimethyl-4-(3-hydroxyphenyl)piperidine], a 3,4-dimethyl-4-(3-hydroxyphenyl)-piperidine, was an inverse agonist at the mu-opioid receptor, and the peptide antagonist CTAP (H-D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH(2)) showed variable, assay-dependent properties. All the antagonists precipitated the same degree of cAMP overshoot in opioid-dependent cells. Antagonists at the mu-opioid receptor may be neutral or show inverse agonist activity. Formation of a constitutively active mu-opioid receptor is not a requirement for the development or expression of adenylyl cyclase sensitization.
Product Description
Supelco
6β-Naltrexol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution