Merck
  • Enzymatic removal of mannose moieties can increase the immune response to HIV-1 gp120 in vivo.
Enzymatic removal of mannose moieties can increase the immune response to HIV-1 gp120 in vivo.
Virology (2009-05-05)
Kaustuv Banerjee, Sofija Andjelic, Per Johan Klasse, Yun Kang, Rogier W Sanders, Elizabeth Michael, Robert J Durso, Thomas J Ketas, William C Olson, John P Moore
ABSTRACT
The Env glycoproteins gp120 and gp41 are used in humoral immunity-based vaccines against human immunodeficiency virus (HIV-1) infection. One among many obstacles to such a vaccine is the structural defenses of Env glycoproteins that limit their immunogenicity. For example, gp120 mannose residues can induce immunosuppressive responses in vitro, including IL-10 expression, via mannose C-type lectin receptors on antigen-presenting cells. Here, we have investigated whether mannose removal alters gp120 immunogenicity in mice. Administering demannosylated gp120 (D-gp120) in the T(H)2-skewing adjuvant Alum induced approximately 50-fold higher titers of anti-gp120 IgG, compared to unmodified gp120. While the IgG subclass profile was predominantly T(H)2-associated IgG1, Abs of the T(H)1-associated IgG2a and IgG3 subclasses were also detectable in D-gp120 recipients. Immunizing with D-gp120 also improved T-cell responses. Giving an IL-10 receptor blocking MAb together with unmodified gp120 in Alum increased the anti-gp120 IgG titer, implicating IL-10 as a possible mediator of auto-suppressive responses to gp120.
Sigma-Aldrich
Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade