Merck
  • Amphiphilic pig intestinal microvillus maltase/glucoamylase. Structure and specificity.
Amphiphilic pig intestinal microvillus maltase/glucoamylase. Structure and specificity.
European journal of biochemistry (1982-09-01)
S H Sørensen, O Norén, H Sjöström, E M Danielsen
Sigma-Aldrich
α-Glucosidase from Bacillus stearothermophilus, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
α-Glucosidase from rice, Type V, ammonium sulfate suspension, 40-80 units/mg protein