Merck
  • On-line gas chromatography combustion/pyrolysis isotope ratio mass spectrometry (HRGC-C/P-IRMS) of pineapple (Ananas comosus L. Merr.) volatiles.
On-line gas chromatography combustion/pyrolysis isotope ratio mass spectrometry (HRGC-C/P-IRMS) of pineapple (Ananas comosus L. Merr.) volatiles.
Journal of agricultural and food chemistry (2003-12-24)
Christina Preston, Elke Richling, Sandra Elss, Markus Appel, Frank Heckel, Ariane Hartlieb, Peter Schreier
ABSTRACT
By use of extracts prepared by liquid-liquid separation of the volatiles from self-prepared juices of pineapple fruits (Ananas comosus) (n = 14) as well as commercial pineapple recovery aromas/water phases (n = 3), on-line capillary gas chromatography-isotope ratio mass spectrometry was employed in the combustion (C) and the pyrolysis (P) modes (HRGC-C/P-IRMS) to determine the delta(13)C(VPDB) and delta(2)H(VSMOW) values of selected pineapple flavor constituents. In addition to methyl 2-methylbutanoate 1, ethyl 2-methylbutanoate 2, methyl hexanoate 3, ethyl hexanoate 4, and 2,5-dimethyl-4-methoxy-3[2H]-furanone 5, each originating from the fruit, the delta(13)C(VPDB) and delta(2)H(VSMOW) data of commercial synthetic 1-5 and "natural" (biotechnologically derived) 1-4 were determined. With delta(13)C(VPDB) data of pineapple volatiles 1-4 varying from -12.8 to -24.4 per thousand, the range expected for CAM metabolism was observed. Compound 5 showed higher depletion from -20.9 to -28.6 per thousand. A similar situation was given for the delta(2)H(VSMOW) values of 3-5 from pineapple ranging from -118 to -191 per thousand, whereas 1 and 2 showed higher depleted values from -184 to -263 per thousand. In nearly all cases, analytical differentiation of 1-5 from pineapple and natural as well as synthetic origin was possible. In general, natural and synthetic 1-5 exhibited delta(13)C(VPDB) data ranging from -11.8 to -32.2 per thousand and -22.7 to -35.9 per thousand, respectively. Their delta(2)H(VSMOW) data were in the range from -242 to -323 per thousand and -49 to -163 per thousand, respectively.
Sigma-Aldrich
Ethyl hexanoate, ≥99%
Sigma-Aldrich
Ethyl hexanoate, natural, ≥98%, FCC, FG
Supelco
Methyl hexanoate, analytical standard
Sigma-Aldrich
Methyl hexanoate, natural, ≥99%, FG
Sigma-Aldrich
Methyl hexanoate, ≥99%, FG
Sigma-Aldrich
Methyl hexanoate, 99%
Sigma-Aldrich
Ethyl hexanoate, ≥98%, FCC, FG