Merck
Preparation of guanine and diaminopurine from biuret. Part III.
Chemistry & biodiversity (2007-04-20)
Irene M Lagoja, Piet Herdewijn
ABSTRACT
Because of their potential prebiotic origin and relative chemical stability, urea, biuret, formic acid, and glycine amide might have played a role in the assembly process of purine bases. In this paper, we describe a short reaction path to purine nucleobases from these acyclic precursors. The formation of different purines was verified by UV and NMR spectroscopy, as well as by mass spectrometry.
Sigma-Aldrich
Glycinamide hydrochloride, 98%
Sigma-Aldrich
Glycinamide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Biuret, 98% (Contains ≤10%(w/w) Triuret)