Merck
Bioconversion of heptanal to heptanol by Saccharomyces cerevisiae.
Yeast (Chichester, England) (2010-02-02)
Shikha Verma, Aurobindo Kumar Ray, Bijay Krishna De
ABSTRACT
Saccharomyces cerevisiae is widely known for its catalytic activity on substrates such as aldehyde and ketone. Interestingly, the activity of S. cerevisiae on heptanal (C(6)H(13)CHO), in spite of its being a very common aldehyde, has not been explored. The main objective of this study was therefore to investigate the bioconversion of heptanal, using a strain of the yeast S. cerevisiae. Bioconversion parameters such as incubation period, pH, concentration of substrate, yeast and maltose were also optimized. The study revealed heptanol as the major product. The optimum conditions for biotransformation were found to be: 3 days incubation; pH 7.0; heptanal concentration 0.15 ml/100 ml medium; and S. cerevisiae concentration of 0.15 g/100 ml medium. Reduction in maltose content (to 0.3 g maltose/100 ml medium) showed increased conversion of heptanal. Heptanoic acid and 2-hydroxyheptanoic acid were obtained as two minor co-products. The overall study showed that S. cerevisiae converted heptanal to heptanol by a yield of 68.9 +/- 1.1% w/w under optimum conditions.
Sigma-Aldrich
1-Heptanol, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Heptanol, 98%
Pricing and availability is not currently available.
Supelco
1-Heptanol, analytical standard, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Heptyl alcohol, ≥97%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Heptaldehyde, 95%
Pricing and availability is not currently available.