Merck
  • The mimic of type II aldolases chemistry: asymmetric synthesis of beta-hydroxy ketones by direct aldol reaction.
The mimic of type II aldolases chemistry: asymmetric synthesis of beta-hydroxy ketones by direct aldol reaction.
Chemical biology & drug design (2010-06-25)
Zhijin Lu, Haibo Mei, Jianlin Han, Yi Pan
ABSTRACT
An efficient direct aldol reaction has been developed for the synthesis of chiral beta-hydroxy ketone using a combination of C(1)-symmetric chiral prolinamides based on o-phenylenediamine and zinc triflate as catalyst. The reaction was convenient to carry out in aqueous media with up to 98% chemical yields and up to 94% ee values. The current strategy can be regarded as the analogue of aldolase type II, which suggests a new pathway for the designing of new organocatalysts.
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
L-Prolinamide, 98%
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%