- Search for influence of spatial properties on affinity at α1-adrenoceptor subtypes for phenylpiperazine derivatives of phenytoin.
Search for influence of spatial properties on affinity at α1-adrenoceptor subtypes for phenylpiperazine derivatives of phenytoin.
Bioorganic & medicinal chemistry letters (2010-09-04)
Jadwiga Handzlik, Heinz H Pertz, Tilo Görnemann, Sven Jähnichen, Katarzyna Kieć-Kononowicz
PMID20813529
ABSTRACT
A series of phenylpiperazine derivatives of phenytoin was evaluated for their affinity at α(1)-adrenoceptor subtypes in functional bioassays (rat tail artery: α(1A) and/or α(1B); guinea pig spleen: α(1B); rat aorta: α(1D)). The most potent compounds at α(1A)-, α(1B)- and α(1D)-adrenoceptors, 11, 18 and 8, showed affinities in the submicromolar range. The role of a hydrogen bond donor group for affinity and selectivity at α(1B)-adrenoceptors, postulated by Bremner's pharmacophore model, was confirmed by functional and molecular modelling studies.