Merck
New idea of a small-sized neutron detector with a plastic fibre.
Radiation protection dosimetry (2011-04-29)
Tetsuro Matsumoto, Hideki Harano, Akihiko Masuda, Jun Nishiyama, Yoshinori Sakurai, Akira Uritani
ABSTRACT
A small-sized thermal neutron detector based on a (6)Li-glass scintillator and a plastic optical fibre has been developed for precise measurement of the spatial distribution of thermal neutron fluence rate. This detector was tested in experiments performed using thermal neutrons. The detector is useful in a thermal neutron field. However, the gamma-ray discrimination ability of the (6)Li-glass scintillator is not good in a low-intensity thermal neutron field. A new idea using a gamma-ray suppression method is proposed to reduce the uncertainty in the neutron counts due to the gamma-ray background. A novel small-sized thermal neutron detector consists of a (6)Li-glass scintillator, a hollow CsI(Tl) scintillator and plastic optical fibres. The evaluation of the gamma-ray suppression ability of the detector using the EGS4 code indicates that the gamma-ray suppression is effective.
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis
Sigma-Aldrich
Cesium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Cesium iodide, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Cesium iodide, 99.999% trace metals basis