Merck
  • Expression and characterization of an extremely thermostable β-glycosidase (mannosidase) from the hyperthermophilic archaeon Pyrococcus furiosus DSM3638.
Expression and characterization of an extremely thermostable β-glycosidase (mannosidase) from the hyperthermophilic archaeon Pyrococcus furiosus DSM3638.
New biotechnology (2011-06-01)
Sung H Park, Kwan H Park, Byung C Oh, Inteaz Alli, Byong H Lee
ABSTRACT
Genomic analysis of the hyperthermophilic archaeon Pyrococcus furiosus revealed the presence of an open reading frame (ORF PF0356) similar to the enzymes in glycoside hydrolase family 1. This β-glycosidase, designated PFTG (P. furiosus thermostable glycosidase), was cloned and expressed in Escherichia coli. The expressed enzyme was purified by heat treatment and Ni-NTA affinity chromatography. The gene was composed of 1,452 bp encoding 483 amino acids for a protein with a predicted molecular mass of 56,326 Da. The temperature and pH optima were 100°C and 5.0 in sodium citrate buffer, respectively. The substrate specificity of PFTG suggests that it possesses characteristics of both β-galactosidase and β-mannosidase activities. However, through kinetic studies by ITC (Isothermal Titration Colorimetry) which is very sensitive method for enzyme kinetics, PF0356 enzyme revealed the highest catalytic efficiency toward p-nitrophenyl-β-d-mannopyranoside (3.02 k(cat)/K(m)) and mannobiose (4.32 k(cat)/K(m)). The enzyme showed transglycosylation and transgalactosylation activities toward cellobiose, lactose and mannooligosaccharides that could produce GOS (galactooligosaccharides) and MOS (maltooligosaccharides). This novel hyperthermostable β-glycosidase may be useful for food and pharmaceutical applications.
Sigma-Aldrich
4-Nitrophenyl β-D-mannopyranoside, ≥98%
Pricing and availability is not currently available.