Merck
  • Amino acid metabolism in the human fetus at term: leucine, valine, and methionine kinetics.
Amino acid metabolism in the human fetus at term: leucine, valine, and methionine kinetics.
Pediatric research (2011-08-23)
Chris H P van den Akker, Henk Schierbeek, Gardi Minderman, Andras Vermes, Ernst M Schoonderwaldt, Johannes J Duvekot, Eric A P Steegers, Johannes B van Goudoever
ABSTRACT
Human fetal metabolism is largely unexplored. Understanding how a healthy fetus achieves its fast growth rates could eventually play a pivotal role in improving future nutritional strategies for premature infants. To quantify specific fetal amino acid kinetics, eight healthy pregnant women received before elective cesarean section at term, continuous stable isotope infusions of the essential amino acids [1-13C,15N]leucine, [U-13C5]valine, and [1-13C]methionine. Umbilical blood was collected after birth and analyzed for enrichments and concentrations using mass spectrometry techniques. Fetuses showed considerable leucine, valine, and methionine uptake and high turnover rates. α-Ketoisocaproate, but not α-ketoisovalerate (the leucine and valine ketoacids, respectively), was transported at net rate from the fetus to the placenta. Especially, leucine and valine data suggested high oxidation rates, up to half of net uptake. This was supported by relatively low α-ketoisocaproate reamination rates to leucine. Our data suggest high protein breakdown and synthesis rates, comparable with, or even slightly higher than in premature infants. The relatively large uptakes of total leucine and valine carbon also suggest high fetal oxidation rates of these essential branched chain amino acids.
Sigma-Aldrich
4-Methyl-2-oxovaleric acid, ≥98.0% (T)
Sigma-Aldrich
Sodium 4-methyl-2-oxovalerate, leucine metabolite
Sigma-Aldrich
Sodium 3-methyl-2-oxobutyrate, 95%